Pyrazine-2-carboxylic acid, 99%, Thermo Scientific Chemicals
Pyrazine-2-carboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyrazine-2-carboxylic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A13363.14
also known as A13363-14
Price (EUR)/ Each
28,44
Online exclusive
31,60 
Save 3,16 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)/ Each
28,44
Online exclusive
31,60 
Save 3,16 (10%)
Add to cart
Pyrazine-2-carboxylic acid, 99%, Thermo Scientific Chemicals
Catalog numberA13363.14
Price (EUR)/ Each
28,44
Online exclusive
31,60 
Save 3,16 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namepyrazine-2-carboxylic acid
Molecular FormulaC5H4N2O2
InChI KeyNIPZZXUFJPQHNH-UHFFFAOYSA-N
SMILESOC(=O)C1=CN=CC=N1
Molecular Weight (g/mol)124.10
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only