Thermo Scientific Chemicals

4-n-Hexyloxybenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A13367.22
100 g, Each
Thermo Scientific Chemicals

4-n-Hexyloxybenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A13367.22
100 g, Each
Quantity
Catalog number: A13367.22
also known as A13367-22
Price (EUR)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39649-71-3
IUPAC Name
4-(hexyloxy)benzoyl chloride
Molecular Formula
C13H17ClO2
InChI Key
DQQOONVCLQZWOY-UHFFFAOYSA-N
SMILES
CCCCCCOC1=CC=C(C=C1)C(Cl)=O
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Refractive Index
1.5355-1.5395 @ 20?C
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    EU Specific Hazard Statements

    EUH029 - Contact with water liberates toxic gas

    Precautionary Statements

    P402 + P404 - Store in a dry place. Store in a closed container

    P280 - Wear eye protection/ face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician