Thermo Scientific Chemicals

Dimethyl phthalate, 99%, Thermo Scientific Chemicals

Catalog number: A14253.30
250 g, Each
Thermo Scientific Chemicals

Dimethyl phthalate, 99%, Thermo Scientific Chemicals

Catalog number: A14253.30
250 g, Each
Quantity
Catalog number: A14253.30
also known as A14253-30
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
131-11-3
IUPAC Name
1,2-dimethyl benzene-1,2-dicarboxylate
Molecular Formula
C10H10O4
InChI Key
NIQCNGHVCWTJSM-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=C1C(=O)OC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.5140-1.5200 @ 20?C

Description

Dimethyl phthalate is used as a plasticizer, solvent, rubber coating agent, molding powder, finishing agent and intermediate. Further, it is used in solid rocket propellants, safety glass, lacquer coatings, adhesives and sealants, toner and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl phthalate is used as a plasticizer, solvent, rubber coating agent, molding powder, finishing agent and intermediate. Further, it is used in solid rocket propellants, safety glass, lacquer coatings, adhesives and sealants, toner and colorant products.

Solubility
Miscible with alcohol, ethanol, ether, benzene, chloroform, esters, ketones, organic solvents and chlorinated hydrocarbons. Slightly miscible with carbon tetrachloride. Immiscible with water, petroleum ether and paraffin hydrocarbons.

Notes
Incompatible with oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text