Thermo Scientific Chemicals

Creatine phosphate disodium salt tetrahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A15362.14
25 g, Each
Thermo Scientific Chemicals

Creatine phosphate disodium salt tetrahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A15362.14
25 g, Each
Quantity
Catalog number: A15362.14
also known as A15362-14
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
71519-72-7
IUPAC Name
disodium tetrahydrate ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
Molecular Formula
C4H16N3Na2O9P
InChI Key
HUWYWJSJJDCZRQ-UHFFFAOYSA-L
SMILES
O.O.O.O.[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
Appearance (Color)
White
Form
Powder or crystalline powder
Impurity content
Creatine: ≤0.6%
Assay from Supplier's CofA
≥98.0% (HPLC - Dry wt. basis)
Water Content (Karl Fischer Titration)
≥20.0 to ≤25.5% (3.54-4.85 waters)

Description

Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.

Solubility
Soluble in water.0.1 g/mL.

Notes
Incompatible with Strong bases. strong acids. Protect from Direct sunlight.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text