Thermo Scientific Chemicals

4-Chlorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A16325.22
100 g, Each
Thermo Scientific Chemicals

4-Chlorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A16325.22
100 g, Each
Quantity
Catalog number: A16325.22
also known as A16325-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
122-01-0
IUPAC Name
4-chlorobenzoyl chloride
Molecular Formula
C7H4Cl2O
InChI Key
RKIDDEGICSMIJA-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=C(Cl)C=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid (low melting solid)
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Refractive Index
1.5760-1.5800 @ 20?C

Description

4-Chlorobenzoyl chloride was used to synthesize 4-chlorobenzoyl CoA by reacting it with CoA in KHCO3 buffer. 4-Chlorobenzoyl chloride may be used in the following studies Acylation of benzene using different solid acid catalysts such as dodecatungstophosphoric acid (DTPA), DTPA/K-10 clay, K-10, Amberlite, Amberlyst-15, Indion-130, Filtrol-24 clay and sulfated zirconia, Preparation of 1-(4-chlorobenzoyl)-2, 7-dimethoxynaphthalene, Preparation of 4-chlorobenzoyl CoA, via reaction with CoA in KHCO3 buffer, Preparation of 1-aryloxyacetyl-4-(4-chlorobenzoyl)-semicarbazides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzoyl chloride was used to synthesize 4-chlorobenzoyl CoA by reacting it with CoA in KHCO3 buffer. 4-Chlorobenzoyl chloride may be used in the following studies Acylation of benzene using different solid acid catalysts such as dodecatungstophosphoric acid (DTPA), DTPA/K-10 clay, K-10, Amberlite, Amberlyst-15, Indion-130, Filtrol-24 clay and sulfated zirconia, Preparation of 1-(4-chlorobenzoyl)-2, 7-dimethoxynaphthalene, Preparation of 4-chlorobenzoyl CoA, via reaction with CoA in KHCO3 buffer, Preparation of 1-aryloxyacetyl-4-(4-chlorobenzoyl)-semicarbazides.

Solubility
Reacts with water. Reacts with alcohol.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, alcohol, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    H335 - May cause respiratory irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician