Thermo Scientific Chemicals

N-Fmoc-L-alanine, 95%, Thermo Scientific Chemicals

Catalog number: H59438.14
25 g, Each
Thermo Scientific Chemicals

N-Fmoc-L-alanine, 95%, Thermo Scientific Chemicals

Catalog number: H59438.14
25 g, Each
Quantity
Catalog number: H59438.14
also known as H59438-14
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
35661-39-3
IUPAC Name
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Molecular Formula
C18H17NO4
InChI Key
QWXZOFZKSQXPDC-UHFFFAOYNA-N
SMILES
CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Assay (HPLC)
94.0% min

Description

N-Fmoc-L-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Alanine is one of the simplest amino acids - a methyl group as the side chain. This small side chain confers a high degree of flexibility when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Alanine is one of the simplest amino acids - a methyl group as the side chain. This small side chain confers a high degree of flexibility when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text