Thermo Scientific Chemicals

Dicloxacillin sodium salt monohydrate, Thermo Scientific Chemicals

Catalog number: J61581.03
1 g, Each
Thermo Scientific Chemicals

Dicloxacillin sodium salt monohydrate, Thermo Scientific Chemicals

Catalog number: J61581.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61581.03
also known as J61581-03
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
13412-64-1
IUPAC Name
sodium (2S,5R,6R)-6-[3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-amido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrate
Molecular Formula
C19H18Cl2N3NaO6S
InChI Key
SIGZQNJITOWQEF-VICXVTCVSA-M
SMILES
O.[Na+].CC1=C(C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C(=NO1)C1=C(Cl)C=CC=C1Cl
Assay (HPLC)
95 - 102%
Water Content (Karl Fischer Titration)
3.0-5.0%
Appearance (Color)
White to off-white
Form
Powder
Assay (unspecified)
850?g/mg min.

Description

Dicloxacillin sodium salt is used in the treatment of staphylococci resistant to penicillin G. Dicloxacillin binds to specific penicillin-binding proteins in the bacterial cell wall and therefore inhibits the last stage of bacterial cell wall synthesis. Cell lysis, mediated by bacterial cell wall autolytic enzymes, is the result. Dicloxacillin may interfere with autolysin inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Respiratory Sensitization
    Category 1
    Skin Sensitization
    Category 1
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H315 - Causes skin irritation

    H317 - May cause an allergic skin reaction

    H319 - Causes serious eye irritation

    H334 - May cause allergy or asthma symptoms or breathing difficulties if inhaled

    H335 - May cause respiratory irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P332 + P313 - If skin irritation occurs: Get medical advice/attention

    P333 + P313 - If skin irritation or rash occurs: Get medical advice/attention

    P337 + P313 - If eye irritation persists: Get medical advice/attention