Thermo Scientific Chemicals

Carga de plata, 99,999 % (base metálica), Thermo Scientific Chemicals, Premion™

Número de catálogo: 014153.18
50 g, Each
Thermo Scientific Chemicals

Carga de plata, 99,999 % (base metálica), Thermo Scientific Chemicals, Premion™

Número de catálogo: 014153.18
50 g, Each
Cantidad
Número de catálogo: 014153.18
también denominado 014153-18
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
7440-22-4
IUPAC Name
silver
Molecular Formula
Ag
InChI Key
BQCADISMDOOEFD-UHFFFAOYSA-N
SMILES
[Ag]
Appearance (Color)
Silver
Assay from Suppliers CofA
Total metal impurities: ≤0.001% (≤10ppm)
Diameter
90% is ≥1.5mm to ≤3.5mm
Form
Shot

Descripción

For vapor deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text