1,2-Dicloroetano, ACS, +99 %, Thermo Scientific Chemicals
1,2-Dicloroetano, ACS, +99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Dicloroetano, ACS, +99 %, Thermo Scientific Chemicals

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.002%, Titratable acid 0.0003meq/g, H2O 0.03%
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
4 x 1 L
500 mL
1 L
Este producto químico puede requerir que obtengamos información adicional para nuestros registros de cumplimiento normativo y químico. Si es necesario, nos pondremos en contacto con usted para obtener esta información una vez que haya realizado su pedido.
Número de catálogo 039121.M1
también denominado 039121-M1
Precio (EUR)/ Each
209,00
-
Añadir al carro de la compra
Cantidad:
4 x 1 L
Pedido a granel o personalizado
Precio (EUR)/ Each
209,00
Añadir al carro de la compra
1,2-Dicloroetano, ACS, +99 %, Thermo Scientific Chemicals
Número de catálogo039121.M1
Precio (EUR)/ Each
209,00
-
Añadir al carro de la compra
Identificadores químicos
CAS107-06-2
IUPAC Name1,2-dichloroethane
Molecular FormulaC2H4Cl2
InChI KeyWSLDOOZREJYCGB-UHFFFAOYSA-N
SMILESClCCCl
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear colorless
FormLiquid
Color (APHA)≤10
Assay from Suppliers CofA≥99.0%
Free acid (titration)≤0.0003 meq/g
Ver más
Used to make vinyl chloride, and chlorinated solvents(trichloroethane, trichloroethylene, perchloroethylene, and vinylidene), remove grease, resins, glue and dirt. Used as a solvent in the manufacturing of polystyrene and SBR latex. It is added to leaded petrol as an anti-knock compound. It is also used in polyvinyl chloride (PVC) pipes, packaging materials, furniture, auto mobile parts, wall coverings and housewares. It is also used generally as an intermediate for other organic chemical compounds and as a solvent. Used as fumigant residues for wheat, flour, bran, middlings and bread. Used as solvent for the isomer distribution in the acetylation of benzene and toluene under the influence of aluminum chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only