Thermo Scientific Chemicals

Trihidrato de cloramina-T, ACS, 98,0-103,0 %, Thermo Scientific Chemicals

Número de catálogo: 042374.36
500 g, Each
Thermo Scientific Chemicals

Trihidrato de cloramina-T, ACS, 98,0-103,0 %, Thermo Scientific Chemicals

Número de catálogo: 042374.36
500 g, Each
Cantidad
Número de catálogo: 042374.36
también denominado 042374-36
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
7080-50-4
IUPAC Name
sodium (chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate trihydrate
Molecular Formula
C7H13ClNNaO5S
InChI Key
RNROOTWAJBXJBD-UHFFFAOYSA-M
SMILES
O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl
Comment
Suitable for determination of bromide P.T.
Comment
Clarity of aqueous solution P.T.
Impurity content
Insoluble in alcohol 1.5%
pH
of a 5% solution 8.0-10.0 at25?

Descripción

Reagent for selective oxidation of methionineChloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Skin Corrosion/Irritation
    Category 1
    Respiratory Sensitization
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 - Harmful if swallowed

    H314 - Causes severe skin burns and eye damage

    H334 - May cause allergy or asthma symptoms or breathing difficulties if inhaled

    EU Specific Hazard Statements

    EUH031 - Contact with acids liberates toxic gas

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear eye protection/ face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing