Guaiacol, +99 %, Thermo Scientific Chemicals
Guaiacol, +99 %, Thermo Scientific Chemicals
Guaiacol, +99 %, Thermo Scientific Chemicals
Guaiacol, +99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Guaiacol, +99 %, Thermo Scientific Chemicals

CAS: 90-05-1 | C7H8O2 | 124.14 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 kg
250 g
1 kg
Envase:
Ver todo
Tambor de plástico
Número de catálogo 120190050
Precio (EUR)
256,70
Exklusiv online
310,00
Ahorro 53,30 (17%)
Each
-
Añadir al carro de la compra
Cantidad:
5 kg
Envase:
Tambor de plástico
Recurring order eligible. Learn more »
Pedido a granel o personalizado
Precio (EUR)
256,70
Exklusiv online
310,00
Ahorro 53,30 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS90-05-1
IUPAC Name2-methoxyphenol
Molecular FormulaC7H8O2
InChI KeyLHGVFZTZFXWLCP-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid or crystalline mass
Infrared spectrumConforms
GC>=99.0 %
Refractive index1.5420 to 1.5470 (20°C, 589 nm)
Ask our AI about this Product

Este producto de marca Thermo Scientific Chemicals formó parte originalmente de la cartera de productos Acros Organics. Alguna información sobre documentos y etiquetas puede referirse a la marca original. El producto original Acros Organics/código de artículo o referencia SKU no ha cambiado como parte de la transición de marca a Thermo Scientific Chemicals.

RUO - Para uso exclusivo en investigación
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
A0462681Certificate of Analysis17 may 2025120190010, 120190050, 120192500
A0429990Certificate of Analysis17 may 2025120190010, 120190050, 120192500
A0467708Certificate of Analysis30 ene 2025120190010, 120190050, 120192500
A0462681Certificate of Analysis13 may 2024120190010, 120190050, 120192500
A0429990Certificate of Analysis10 may 2024120190010, 120190050, 120192500
Se muestran 5 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Label Elements
Signal Word

Warning

Hazard Statements

H302 - Harmful if swallowed

H315 - Causes skin irritation

H319 - Causes serious eye irritation

EU Specific Hazard Statements

Combustible liquid

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P362 - Take off contaminated clothing and wash before reuse

Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection