Hidroquinona, 99 %, Thermo Scientific Chemicals
Hidroquinona, 99 %, Thermo Scientific Chemicals
Hidroquinona, 99 %, Thermo Scientific Chemicals
Hidroquinona, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hidroquinona, 99 %, Thermo Scientific Chemicals

CAS: 123-31-9 | C6H6O2 | 110.11 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
10 kg
500 g
2 kg
Envase:
Ver todo
Tambor de plástico
Número de catálogo 120910100
Precio (EUR)
389,30
Precio exclusivo en nuestra web
469,00
Ahorro 79,70 (17%)
Each
-
Añadir al carro de la compra
Cantidad:
10 kg
Envase:
Tambor de plástico
Pedido a granel o personalizado
Precio (EUR)
389,30
Precio exclusivo en nuestra web
469,00
Ahorro 79,70 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS123-31-9
IUPAC Namebenzene-1,4-diol
Molecular FormulaC6H6O2
InChI KeyQIGBRXMKCJKVMJ-UHFFFAOYSA-N
SMILESOC1=CC=C(O)C=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to off-white
Melting point170°C to 175°C
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals or needles
HPLC>=98.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Serious Eye Damage/Eye Irritation
Category 1
Skin Sensitization
Category 1
Germ Cell Mutagenicity
Category 2
Carcinogenicity
Category 2
Acute aquatic toxicity
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H302 - Harmful if swallowed

H317 - May cause an allergic skin reaction

H318 - Causes serious eye damage

H341 - Suspected of causing genetic defects

H351 - Suspected of causing cancer

H400 - Very toxic to aquatic life

Precautionary Statements

P273 - Avoid release to the environment

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Compartir número de catálogo, nombre o enlace.