Ácido decanoico, 99 %, Thermo Scientific Chemicals
Ácido decanoico, 99 %, Thermo Scientific Chemicals
Ácido decanoico, 99 %, Thermo Scientific Chemicals
Ácido decanoico, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido decanoico, 99 %, Thermo Scientific Chemicals

CAS: 334-48-5 | C10H20O2 | 172.268 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
500g
1kg
2,5 kg
Envase:
Ver todo
Frasco de vidrio
Número de catálogo 167271000
Precio (EUR)
36,21
Online Exclusive
43,70
Ahorro 7,49 (17%)
Each
-
Añadir al carro de la compra
Cantidad:
100 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
36,21
Online Exclusive
43,70
Ahorro 7,49 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS334-48-5
IUPAC Namedecanoic acid
Molecular FormulaC10H20O2
InChI KeyGHVNFZFCNZKVNT-UHFFFAOYSA-N
SMILESCCCCCCCCCC(O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Crystals and/or chunks or fused solid or liquid
Melting point29°C to 34°C
Infrared spectrumConforms
Appearance (Color)Colorless or white to cream
GC>=98.5 %
Ask our AI about this Product

Este producto de marca Thermo Scientific Chemicals formó parte originalmente de la cartera de productos Acros Organics. Alguna información sobre documentos y etiquetas puede referirse a la marca original. El producto original Acros Organics/código de artículo o referencia SKU no ha cambiado como parte de la transición de marca a Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Chronic aquatic toxicity
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H412 - Harmful to aquatic life with long lasting effects

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P362 - Take off contaminated clothing and wash before reuse

Compartir número de catálogo, nombre o enlace.