2-Naftol, 99+ %, Thermo Scientific Chemicals
2-Naftol, 99+ %, Thermo Scientific Chemicals
2-Naftol, 99+ %, Thermo Scientific Chemicals
2-Naftol, 99+ %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Naftol, 99+ %, Thermo Scientific Chemicals

CAS: 135-19-3 | C10H8O | 144.17 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
500 g
1 kg
2.5 kg
Envase:
Ver todo
Frasco de vidrio
Número de catálogo 180820010
Precio (EUR)
147,05
Online Exclusive
177,00
Ahorro 29,95 (17%)
Each
-
Añadir al carro de la compra
Cantidad:
1 kg
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
147,05
Online Exclusive
177,00
Ahorro 29,95 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS135-19-3
IUPAC Namenaphthalen-2-ol
Molecular FormulaC10H8O
InChI KeyJWAZRIHNYRIHIV-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC=CC2=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
GC>=99.0 %
Appearance (Color)White to beige to pink to light brown
Appearance (Form)Powder or flakes
Infrared spectrumConforms
Melting point120°C to 124°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documentos y descargas

Certificados

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Acute Inhalation Toxicity - Dusts and Mists
Category 4
Acute aquatic toxicity
Category 1
Label Elements
Signal Word

Warning

Hazard Statements

H302 + H332 - Harmful if swallowed or if inhaled

H400 - Very toxic to aquatic life

Precautionary Statements

P273 - Avoid release to the environment

P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

P304 + P340 - IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing

Compartir número de catálogo, nombre o enlace.