Cianato de potasio, 97 %, puro, Thermo Scientific Chemicals
Cianato de potasio, 97 %, puro, Thermo Scientific Chemicals
Cianato de potasio, 97 %, puro, Thermo Scientific Chemicals
Cianato de potasio, 97 %, puro, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cianato de potasio, 97 %, puro, Thermo Scientific Chemicals

CAS: 590-28-3 | CKNO | 81.12 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
100 g
1 kg
Envase:
Frasco de plástico
Número de catálogo 201115000
Precio (EUR)/ Each
46,16
Offre exceptionnelle en ligne
54,30
Ahorro 8,14 (15%)
Añadir al carro de la compra
Cantidad:
500 g
Envase:
Frasco de plástico
Pedido a granel o personalizado
Precio (EUR)/ Each
46,16
Offre exceptionnelle en ligne
54,30
Ahorro 8,14 (15%)
Añadir al carro de la compra
Identificadores químicos
CAS590-28-3
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White
Titration Argentometric>=96.0 %
Appearance (Form)Crystalline powder and/or chunks
Potassium carbonate=<3 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Compartir número de catálogo, nombre o enlace.