1x1 image pixel for data collection
Hidróxido de bario octahidrato, +98 %, para análisis, Thermo Scientific Chemicals
Hidróxido de bario octahidrato, +98 %, para análisis, Thermo Scientific Chemicals
Hidróxido de bario octahidrato, +98 %, para análisis, Thermo Scientific Chemicals
Hidróxido de bario octahidrato, +98 %, para análisis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hidróxido de bario octahidrato, +98 %, para análisis, Thermo Scientific Chemicals

Hidróxido de bario octahidrato, +98 %, para análisis, ACROS Organics, cantidad:1kg, Envasado:Frasco de plástico, CAS:12230-71-6, blanco, grado:análisis, Punto de fusión:78,0 °C, Fórmula molecular:H2BaO2.8H2O, Peso molecular:315,48; Porcentaje de pureza:+98 %
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 kg
250 g
1 kg
Envase:
Frasco de plástico
Número de catálogo 203140050
Precio (EUR)
385,90
Online Exclusive
454,00
Ahorro 68,10 (15%)
Each
Disponibilidad estimada  Pendientes  
Añadir al carro de la compra
Cantidad:
5 kg
Envase:
Frasco de plástico
Pedido a granel o personalizado
Precio (EUR)
385,90
Online Exclusive
454,00
Ahorro 68,10 (15%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS12230-71-6
IUPAC Namebarium(2+) octahydrate dihydroxide
Molecular FormulaBaH18O10
InChI KeyZUDYPQRUOYEARG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Chloride (Cl)=<20 ppm
Sulfur (S)=<5 ppm
Appearance (Form)Crystalline powder or crystals or flakes
Appearance (Color)White
Titration with HCl>=98.0 %
Ver más
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
A0443541Certificate of Analysis31 ene 2025203140010, 203140050, 203142500
A0433846Certificate of Analysis18 mar 2024203140010, 203140050, 203142500
A0432352Certificate of Analysis15 ene 2024203140010, 203140050, 203142500
A0394990Certificate of Analysis14 dic 2022203140010, 203140050, 203142500
A0403043Certificate of Analysis14 dic 2022203140010, 203140050, 203142500
Se muestran 5 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Acute Inhalation Toxicity - Dusts and Mists
Category 4
Skin Corrosion/Irritation
Category 1
Serious Eye Damage/Eye Irritation
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H302 + H332 - Harmful if swallowed or if inhaled

H314 - Causes severe skin burns and eye damage

EU Specific Hazard Statements

EUH071 - Corrosive to the respiratory tract

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection