1x1 image pixel for data collection
Óxido de vanadio(V), 99,6 + %, Thermo Scientific Chemicals
Óxido de vanadio(V), 99,6 + %, Thermo Scientific Chemicals
Óxido de vanadio(V), 99,6 + %, Thermo Scientific Chemicals
Óxido de vanadio(V), 99,6 + %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Óxido de vanadio(V), 99,6 + %, Thermo Scientific Chemicals

CAS: 1314-62-1 | O5V2 | 181.88 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
250 g
1 kg
Envase:
Frasco de vidrio
Número de catálogo 206422500
Precio (EUR)
71,74
Online Exclusive
86,50
Ahorro 14,76 (17%)
Each
Añadir al carro de la compra
Cantidad:
250 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
71,74
Online Exclusive
86,50
Ahorro 14,76 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS1314-62-1
IUPAC Name[(dioxovanadio)oxy]dioxovanadium
Molecular FormulaO5V2
InChI KeyGNTDGMZSJNCJKK-UHFFFAOYSA-N
SMILESO=[V](=O)O[V](=O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Yellow to orange-brown
Potassium (K)=<200 ppm
Iron (Fe)=<300 ppm
Silicon (Si)=<200 ppm
Appearance (Form)Powder
Ver más

Este producto de marca Thermo Scientific Chemicals formó parte originalmente de la cartera de productos Acros Organics. Alguna información sobre documentos y etiquetas puede referirse a la marca original. El producto original Acros Organics/código de artículo o referencia SKU no ha cambiado como parte de la transición de marca a Thermo Scientific Chemicals.

RUO – Research Use Only
Please enter sku to load default questions

Documentos y descargas

Certificados

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 3
Acute Inhalation Toxicity - Dusts and Mists
Category 2
Germ Cell Mutagenicity
Category 2
Carcinogenicity
Category 1B
Reproductive Toxicity
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Specific target organ toxicity - (repeated exposure)
Category 1
Chronic aquatic toxicity
Category 2
Label Elements
Signal Word

Danger

Hazard Statements

H301 - Toxic if swallowed

H330 - Fatal if inhaled

H335 - May cause respiratory irritation

H341 - Suspected of causing genetic defects

H350 - May cause cancer

H361fd - Suspected of damaging fertility. Suspected of damaging the unborn child

H362 - May cause harm to breast-fed children

H372 - Causes damage to organs through prolonged or repeated exposure

H411 - Toxic to aquatic life with long lasting effects

Precautionary Statements

P264 - Wash face, hands and any exposed skin thoroughly after handling

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P310 - Immediately call a POISON CENTER or doctor/physician

Additional EU labelling

Restricted to professional users

Compartir número de catálogo, nombre o enlace.