1x1 image pixel for data collection
Metal de Wood, masa ( 100 mm aprox.), Thermo Scientific Chemicals
Metal de Wood, masa ( 100 mm aprox.), Thermo Scientific Chemicals
Metal de Wood, masa ( 100 mm aprox.), Thermo Scientific Chemicals
Metal de Wood, masa ( 100 mm aprox.), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Metal de Wood, masa ( 100 mm aprox.), Thermo Scientific Chemicals

CAS: 76093-98-6 | Pb·Sn
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
100 g
Envase:
Frasco de vidrio
Número de catálogo 388551000
Precio (EUR)
45,39
Online Exclusive
54,70
Ahorro 9,31 (17%)
Each
Añadir al carro de la compra
Cantidad:
100 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
45,39
Online Exclusive
54,70
Ahorro 9,31 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS76093-98-6
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Silver grey
Compositionalloy typically:Bi 50%; Pb 25%; Cd 12.5%; Sn 12.5%
Appearance (Form)Rods
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Compartir número de catálogo, nombre o enlace.