Clorato de potasio, +99 %, reactivo ACS, Thermo Scientific Chemicals
Clorato de potasio, +99 %, reactivo ACS, Thermo Scientific Chemicals
Clorato de potasio, +99 %, reactivo ACS, Thermo Scientific Chemicals
Clorato de potasio, +99 %, reactivo ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Clorato de potasio, +99 %, reactivo ACS, Thermo Scientific Chemicals

CAS: 3811-04-9 | ClKO3 | 122.55 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
500 g
2.5 kg
Envase:
Ver todo
Frasco de vidrio
Número de catálogo 418190050
Precio (EUR)
20,23
Exklusiv online
24,40
Ahorro 4,17 (17%)
Each
-
Añadir al carro de la compra
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
20,23
Exklusiv online
24,40
Ahorro 4,17 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS3811-04-9
IUPAC Namepotassium chlorate
Molecular FormulaClKO3
InChI KeyVKJKEPKFPUWCAS-UHFFFAOYSA-M
SMILES[K+].[O-][Cl](=O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Heavy metals (as Pb)=<5 ppm
Chloride (Cl)=<10 ppm
Appearance (Form)Crystalline powder
Sulfate (SO4)Passes test (limit about 0.002%)
Iron (Fe)=<3 ppm
Ver más
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reactivo de oxidación
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
A0473218Certificate of Analysis09 jun 2025418190025, 418195000, 418190050
A0417707Certificate of Analysis17 may 2025418190025, 418195000, 418190050
A0465872Certificate of Analysis16 sept 2024418190025, 418195000, 418190050
A0453554Certificate of Analysis25 ago 2023418190025, 418195000, 418190050
A0449452Certificate of Analysis06 feb 2023418190025, 418195000, 418190050
Se muestran 5 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Oxidizing solids
Category 1
Acute oral toxicity
Category 3
Acute Inhalation Toxicity - Dusts and Mists
Category 4
Chronic aquatic toxicity
Category 2
Label Elements
Signal Word

Danger

Hazard Statements

H301 - Toxic if swallowed

H332 - Harmful if inhaled

H411 - Toxic to aquatic life with long lasting effects

Physical hazards

H271 - May cause fire or explosion; strong oxidizer

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P220 - Keep away from clothing and other combustible materials

P264 - Wash face, hands and any exposed skin thoroughly after handling

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P312 - Call a POISON CENTER or doctor if you feel unwell

P371 + P380 + P375 - In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the risk of explosion

Compartir número de catálogo, nombre o enlace.