Etóxido de sodio, 21 % en etanol, AcroSeal™, Thermo Scientific Chemicals
Etóxido de sodio, 21 % en etanol, AcroSeal™, Thermo Scientific Chemicals
Etóxido de sodio, 21 % en etanol, AcroSeal™, Thermo Scientific Chemicals
Etóxido de sodio, 21 % en etanol, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Etóxido de sodio, 21 % en etanol, AcroSeal™, Thermo Scientific Chemicals

CAS: 141-52-6 | C2H5NaO | 68.04 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
800 mL
100 mL
Envase:
Frasco de vidrio AcroSeal™
Número de catálogo 429308000
Precio (EUR)
80,41
Online Exclusive
97,00
Ahorro 16,59 (17%)
Each
-
Añadir al carro de la compra
Cantidad:
800 mL
Envase:
Frasco de vidrio AcroSeal™
Pedido a granel o personalizado
Precio (EUR)
80,41
Online Exclusive
97,00
Ahorro 16,59 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS141-52-6
IUPAC Namesodium ethanolate
Molecular FormulaC2H5NaO
InChI KeyQDRKDTQENPPHOJ-UHFFFAOYSA-N
SMILES[Na+].CC[O-]
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
AppearanceMay darken on storage
Titration with HCl20.0 to 22.0 %
NaOH + Na2CO3 (as NaOH)=<0.5 %
Appearance (Form)Solution
Appearance (Color)Yellow to brown
Ask our AI about this Product

Este producto de marca Thermo Scientific Chemicals formó parte originalmente de la cartera de productos Acros Organics. Alguna información sobre documentos y etiquetas puede referirse a la marca original. El producto original Acros Organics/código de artículo o referencia SKU no ha cambiado como parte de la transición de marca a Thermo Scientific Chemicals.

Indicado para fabricación y uso en el laboratorio. No indicado para uso en bebidas

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable liquids
Category 3
Skin Corrosion/Irritation
Category 1
Serious Eye Damage/Eye Irritation
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H314 - Causes severe skin burns and eye damage

Physical Hazards

H226 - Flammable liquid and vapor

EU Specific Hazard Statements

EUH014 - Reacts violently with water

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Compartir número de catálogo, nombre o enlace.