Clortalidona, 98 %, Thermo Scientific Chemicals
Clortalidona, 98 %, Thermo Scientific Chemicals
Clortalidona, 98 %, Thermo Scientific Chemicals
Clortalidona, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Clortalidona, 98 %, Thermo Scientific Chemicals

CAS: 77-36-1 | C14H11ClN2O4S | 338.77 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
250 mg
1 g
Número de catálogo 460930010
Precio (EUR)
383,35
Exklusiv online
462,00
Ahorro 78,65 (17%)
Each
-
Añadir al carro de la compra
Cantidad:
1 g
Pedido a granel o personalizado
Precio (EUR)
383,35
Exklusiv online
462,00
Ahorro 78,65 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS77-36-1
IUPAC Name2-chloro-5-(1-hydroxy-3-oxo-2,3-dihydro-1H-isoindol-1-yl)benzene-1-sulfonamide
Molecular FormulaC14H11ClN2O4S
InChI KeyJIVPVXMEBJLZRO-UHFFFAOYNA-N
SMILESNS(=O)(=O)C1=CC(=CC=C1Cl)C1(O)NC(=O)C2=CC=CC=C12
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Powder
Loss on drying=<0.5 % (1 g, 105°C)
HPLC>=97.5 %
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
Ask our AI about this Product

Este producto de marca Thermo Scientific Chemicals formó parte originalmente de la cartera de productos Acros Organics. Alguna información sobre documentos y etiquetas puede referirse a la marca original. El producto original Acros Organics/código de artículo o referencia SKU no ha cambiado como parte de la transición de marca a Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
A0443073Certificate of Analysis25 ene 2025460932500, 460930010
A0465797Certificate of Analysis04 sept 2024460932500, 460930010
A0413600Certificate of Analysis13 dic 2022460932500, 460930010
A0443073Certificate of Analysis24 jun 2022460932500, 460930010
A0374234Certificate of Analysis02 ago 2021460932500, 460930010
Se muestran 5 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P312 - Call a POISON CENTER or doctor if you feel unwell

Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection