Thermo Scientific Chemicals

Nickel(II) formate dihydrate, Thermo Scientific Chemicals

Catalog number: 013801.36
500 g, Each
Thermo Scientific Chemicals

Nickel(II) formate dihydrate, Thermo Scientific Chemicals

Catalog number: 013801.36
500 g, Each
Quantity
Catalog number: 013801.36
also known as 013801-36
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
15694-70-9
Assay (unspecified)
Ni 31% min

Description

Nickel(II) formate dihydrate is used in nickel catalyst preparations. It is also used as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Respiratory Sensitization
    Category 1
    Skin Sensitization
    Category 1
    Germ Cell Mutagenicity
    Category 2
    Carcinogenicity
    Category 1B
    Reproductive Toxicity
    Category 1A
    Specific target organ toxicity - (repeated exposure)
    Category 1
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H317 - May cause an allergic skin reaction

    H334 - May cause allergy or asthma symptoms or breathing difficulties if inhaled

    H341 - Suspected of causing genetic defects

    H350 - May cause cancer

    H360 - May damage fertility or the unborn child

    H372 - Causes damage to organs through prolonged or repeated exposure

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P201 - Obtain special instructions before use

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P285 - In case of inadequate ventilation wear respiratory protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P308 + P313 - IF exposed or concerned: Get medical advice/attention

    P342 + P311 - If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician

    Additional EU labelling

    Restricted to professional users