Water, ultrapure, HPLC Grade, Thermo Scientific Chemicals
Water, ultrapure, HPLC Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Water, ultrapure, HPLC Grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
4 x 4 L
Catalog number 022934.K7
Price (EUR)/ Each
98,80
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (EUR)/ Each
98,80
Add to cart
Water, ultrapure, HPLC Grade, Thermo Scientific Chemicals
Catalog number022934.K7
Price (EUR)/ Each
98,80
-
Add to cart
Chemical Identifiers
CAS7732-18-5
IUPAC Namewater
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (unspecified)Specific Conductance at 25 deg C and time packaging: 1.5 microS/cm, max
Optical Absorbance at400 nm 0.005 AU, max- at 300 nm 0.005 AU, max- at 250 nm 0.005 AU, max- at 200 nm 0.010 AU, max- at 190 nm 0.010 AU, max
Residue on Evaporation2 ppm, Max
FormLiquid
Water ultrapure is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry. It is also used in conductometric titration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only