Thermo Scientific Chemicals

Trimethyl phosphite, 97%, Thermo Scientific Chemicals

Catalog number: 030132.A4
2.5 kg, Each
Thermo Scientific Chemicals

Trimethyl phosphite, 97%, Thermo Scientific Chemicals

Catalog number: 030132.A4
2.5 kg, Each
Quantity

Chemical Identifiers

CAS
121-45-9
IUPAC Name
trimethyl phosphite
Molecular Formula
C3H9O3P
InChI Key
CYTQBVOFDCPGCX-UHFFFAOYSA-N
SMILES
COP(OC)OC
Appearance (Color)
Colorless
Form
Liquid
Assay (GC)
≥96.0%

Description

It is an important raw material and intermediate used in Organic synthesis..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Vapors
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P280 - Wear protective gloves/protective clothing

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell