Thermo Scientific Chemicals

Hexanes, mixed isomers, ACS, 98.5+%, Thermo Scientific Chemicals

Catalog number: 033321.K4
2.5 L, Each
Thermo Scientific Chemicals

Hexanes, mixed isomers, ACS, 98.5+%, Thermo Scientific Chemicals

Catalog number: 033321.K4
2.5 L, Each
Quantity
Catalog number: 033321.K4
also known as 033321-K4
Price (EUR)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92112-69-1
IUPAC Name
hexane
Molecular Formula
C6H14
InChI Key
VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES
CCCCCC
Free acid (titration)
(Water soluble) ≤0.0003 meq/g
Form
Liquid
Color (APHA)
≤10
Assay (GC)
≥98.5% hexanes ( sum of 5 isomers, total hexanes, plus methylcyclopentane
Impurity content
Sulfur compounds (as S) ≤0.005%

Description

Hexane mixed isomers is used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds namely butyl lithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 2
    Skin Corrosion/Irritation
    Category 2
    Reproductive Toxicity
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Specific target organ toxicity - (repeated exposure)
    Category 2
    Aspiration Toxicity
    Category 1
    Chronic aquatic toxicity
    Category 2
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H304 - May be fatal if swallowed and enters airways

    H315 - Causes skin irritation

    H336 - May cause drowsiness or dizziness

    H361f - Suspected of damaging fertility

    H373 - May cause damage to organs through prolonged or repeated exposure

    H411 - Toxic to aquatic life with long lasting effects

    Physical Hazards

    H225 - Highly flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P308 + P313 - IF exposed or concerned: Get medical advice/attention