Thermo Scientific Chemicals

Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%, Thermo Scientific Chemicals

Catalog number: 036526.22
100 g, Each
Thermo Scientific Chemicals

Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%, Thermo Scientific Chemicals

Catalog number: 036526.22
100 g, Each
Quantity
Catalog number: 036526.22
also known as 036526-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
13446-34-9
IUPAC Name
manganese(2+) tetrahydrate dichloride
Molecular Formula
Cl2H8MnO4
InChI Key
CNFDGXZLMLFIJV-UHFFFAOYSA-L
SMILES
O.O.O.O.[Cl-].[Cl-].[Mn++]
Impurity content
SO₄ 0.005%
Impurity content
Na 0.05%
Assay (unspecified)
98.0-101.0% Cl₂Mn.4H₂O
Impurity content
Ca 0.005%
Impurity content
Fe 5ppm

Description

It is used in the production of dry cell batteries. It is a precursor material for antiknock compound methylcyclopentadienyl manganese tricarbony. It is also useful for coupling reactions as well as organic synthesis. It is a catalyst for fast and slow dehydrogenation of ball milled lithium alanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Serious Eye Damage/Eye Irritation
    Category 1
    Specific target organ toxicity - (repeated exposure)
    Category 2
    Chronic aquatic toxicity
    Category 2
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 - Toxic if swallowed

    H318 - Causes serious eye damage

    H373 - May cause damage to organs through prolonged or repeated exposure

    H411 - Toxic to aquatic life with long lasting effects

    Precautionary Statements

    P280 - Wear eye protection/ face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician