Glassy carbon splinter powder, 80 to 200μm, type 2, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glassy carbon splinter powder, 80 to 200μm, type 2, Thermo Scientific Chemicals
Used as an electrode material in electrochemistry. It is used to make carbon paste electrodes in electrochemistry.
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 038013.18
also known as 038013-18
Price (EUR)/ Each
130,83
Online exclusive
147.00 
Save 16,17 (11%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)/ Each
130,83
Online exclusive
147.00 
Save 16,17 (11%)
Add to cart
Glassy carbon splinter powder, 80 to 200μm, type 2, Thermo Scientific Chemicals
Catalog number038013.18
Price (EUR)/ Each
130,83
Online exclusive
147.00 
Save 16,17 (11%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA>99.9%
Appearance (Color)Black
FormSplinter powder, 80-200 micron (Glassy Carbon type-2, maximimum service temperature 3000?C).
Glassy carbon powder is widely used as an electrode material in electrochemistry. It is used to make carbon paste electrodes in electrochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only