Glassy carbon plate, 2mm (0.08in) thick, type 2, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glassy carbon plate, 2mm (0.08in) thick, type 2, Thermo Scientific Chemicals
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
50 x 50 mm
Catalog number 038022.GH
also known as 038022-GH
Price (EUR)/ Each
1 283,40
Online exclusive
1426.00 
Save 142,60 (10%)
-
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (EUR)/ Each
1 283,40
Online exclusive
1426.00 
Save 142,60 (10%)
Add to cart
Glassy carbon plate, 2mm (0.08in) thick, type 2, Thermo Scientific Chemicals
Catalog number038022.GH
Price (EUR)/ Each
1 283,40
Online exclusive
1426.00 
Save 142,60 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPlate : 50x50 mm
Appearance (Color)Black
Thickness~2 mm
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only