Thermo Scientific Chemicals

Rhodium(III) sulfate tetrahydrate, Premion™, 99.99% (metals basis), Rh 35.9% min, Thermo Scientific Chemicals

Catalog number: 041016.06
5 g, Each
Thermo Scientific Chemicals

Rhodium(III) sulfate tetrahydrate, Premion™, 99.99% (metals basis), Rh 35.9% min, Thermo Scientific Chemicals

Catalog number: 041016.06
5 g, Each
Quantity
Catalog number: 041016.06
also known as 041016-06
Price (EUR)

Chemical Identifiers

CAS
15274-78-9
IUPAC Name
dirhodium(3+) tetrahydrate trisulfate
Molecular Formula
H8O16Rh2S3
InChI Key
JJITWKYISWZJBI-UHFFFAOYSA-H
SMILES
O.O.O.O.[Rh+3].[Rh+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Total Metal Impurities
0.01% max., Rh 35.9% min

Description

Rhodium(III) sulfate is used as plating reagents and additives for noble metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Substances/mixtures corrosive to metal
    Category 1
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    Physical hazards

    H290 - May be corrosive to metals

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician