Bismuth(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Bismuth(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Catalog number 042863.A1
also known as 042863-A1
Price (EUR)/ Each
991,80
Online exclusive
1102.00 
Save 110,20 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (EUR)/ Each
991,80
Online exclusive
1102.00 
Save 110,20 (10%)
Add to cart
Bismuth(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number042863.A1
Price (EUR)/ Each
991,80
Online exclusive
1102.00 
Save 110,20 (10%)
-
Add to cart
Chemical Identifiers
CAS1304-76-3
IUPAC Namedibismuth(3+) trioxidandiide
Molecular FormulaBi2O3
InChI KeyTYIXMATWDRGMPF-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Bi+3].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Bismuth(III) oxide is used in solid-oxide fuel cells. It is also used in optical glasses, to replace lead oxide in whitewares, luxes, varistor formulations and ceramic capacitor formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only