Thermo Scientific Chemicals

Titanium(II) oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 077126.36
500 g, Each
Thermo Scientific Chemicals

Titanium(II) oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 077126.36
500 g, Each
Quantity
Catalog number: 077126.36
also known as 077126-36
Price (EUR)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12137-20-1
IUPAC Name
oxotitanium
Molecular Formula
OTi
InChI Key
OGIDPMRJRNCKJF-UHFFFAOYSA-N
SMILES
O=[Ti]
Total Metal Impurities
0.5% max.

Description

Titanium(II) oxide associated with vanadium oxide forms a transition-metal-oxide monolayer catalyst, which is used in the selective oxidation and ammoxidation of alkylaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text