Carbol Fuchsin, pure
Carbol Fuchsin, pure
Carbol Fuchsin, pure
Carbol Fuchsin, pure
Thermo Scientific Chemicals

Carbol Fuchsin, pure

CAS: 4197-24-4 | C21H22ClN3 | 351.878 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 195790050
Price (EUR)
37,06
Online Exclusive
44,70
Save 7,64 (17%)
Each
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
37,06
Online Exclusive
44,70
Save 7,64 (17%)
Each
Add to cart
Chemical Identifiers
CAS4197-24-4
IUPAC Namehydrogen 4-[(4-aminophenyl)[(1Z)-4-imino-3-methylcyclohexa-2,5-dien-1-ylidene]methyl]-2-methylaniline chloride
Molecular FormulaC21H22ClN3
InChI KeyHZLHRDBTVSZCBS-GHTYLULLSA-N
SMILES[H+].[Cl-].CC1=CC(=CC=C1N)C(\C1=CC=C(N)C=C1)=C1\C=CC(=N)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
UV spectrumComplies
UV Lambda max(in water) about 542 nm
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Suitability(for microscopy) Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link