Thermo Scientific Chemicals

4-Methoxyphenyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: 218760250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Methoxyphenyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: 218760250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 218760250
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
5416-93-3
IUPAC Name
1-isocyanato-4-methoxybenzene
Molecular Formula
C8H7NO2
InChI Key
FMDGXCSMDZMDHZ-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)N=C=O
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
GC
>=98.5 %
Refractive index
1.5450 to 1.5470 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing