Phosphorus, red, 97%, pure, powder
Phosphorus, red, 97%, pure, powder
Thermo Scientific Chemicals

Phosphorus, red, 97%, pure, powder

CAS: 7723-14-0 | P | 30.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 345975000
Price (EUR)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7723-14-0
IUPAC Namephosphorus
Molecular FormulaP
InChI KeyOAICVXFJPJFONN-UHFFFAOYSA-N
SMILES[P]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Assay>=96.0 % (P)
Appearance (Color)Bordeaux to dark brown
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473110Certificate of AnalysisMay 28, 2025345975000, 345970025
A0470980Certificate of AnalysisMay 17, 2025345975000, 345970025
A0466193Certificate of AnalysisSep 19, 2024345975000, 345970025
A0459520Certificate of AnalysisJan 24, 2024345975000, 345970025
A0455959Certificate of AnalysisAug 29, 2023345975000, 345970025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable solids
Category 1
Chronic aquatic toxicity
Category 3
Label Elements
Signal Word

Danger

Hazard Statements

H228 - Flammable solid

H412 - Harmful to aquatic life with long lasting effects

Precautionary Statements

P233 - Keep container tightly closed

P273 - Avoid release to the environment

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P280 - Wear protective gloves/protective clothing/eye protection/face protection

Share catalog number, name or link

1x1 image pixel for data collection