4,4'-Azodianiline, 95%
4,4'-Azodianiline, 95%
4,4'-Azodianiline, 95%
4,4'-Azodianiline, 95%
Thermo Scientific Chemicals

4,4'-Azodianiline, 95%

CAS: 538-41-0 | C12H12N4 | 212.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 401580250
Price (EUR)
718,25
Online Exclusive
866,00
Save 147,75 (17%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
718,25
Online Exclusive
866,00
Save 147,75 (17%)
Each
Add to cart
Chemical Identifiers
CAS538-41-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
HPLC>=94 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0438419Certificate of AnalysisFeb 11, 2025401580250, 401580050
A0437463Certificate of AnalysisAug 25, 2023401580250, 401580050
A0436832Certificate of AnalysisAug 25, 2023401580250, 401580050
A0438419Certificate of AnalysisAug 25, 2023401580250, 401580050
A0431419Certificate of AnalysisJan 05, 2023401580250, 401580050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 3
Label Elements
Signal Word

Danger

Hazard Statements

H301 - Toxic if swallowed

Precautionary Statements

P264 - Wash face, hands and any exposed skin thoroughly after handling

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P405 - Store locked up

Share catalog number, name or link

1x1 image pixel for data collection