Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Description | For Use With (Equipment) |
---|---|---|
452000TS also known as 452000 | Dual-stack benchtop shaker kit; Includes kneewell side mount and orbital shaker stand | MaxQ™ HP 420 Shaker |
191494 | Four drawer assembly | MaxQ™ HP 420 Shaker |
191591 | Roller base for drawer assembly | MaxQ™ HP 420 Shaker |
194033 | Removable shelf | MaxQ HP 435, 436, 480, 481 |
194040 | Quick change platform adapter | MaxQ HP 416, 430, 435, 436, 480, 481 |
194045 | Gassing manifold (one gas) | MaxQ HP 420, 435, 436, 480, 481 |
194139 | Magnote wipe-off magnet and pen kit (floor models) | MaxQ™ HP 435, 436, 480, 481 Shaker |
194140 | Magnote wipe-off magnet and pen kit (benchtop models) | MaxQ™ HP 416, 430, 420 Shaker |
201113TS also known as 201113 | 6 in. seven-day chart recorder | MaxQ™ HP 435, 436, 480, 481 Shaker |
452001TS also known as 452001 | Orbital shaker stand | MaxQ HP 420 |
452002TS also known as 452002 | Kneewell slide mount, shaker slides out on rails for easy access | MaxQ HP 420 |
760164 | Replacement HEPA filter | MaxQ HP 435, 436, 480, 481, 8000 |
201126 | 7 Day Recorder, 115V | MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000 |
201127 | 7 Day Recorder, 220V | MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000 |
760440 | Replacement HEPA filter | MaxQ HP 435, 436, 480, 481, 8000 |
Get maximum flexibility, regardless of your application, by using Thermo Scientific™ Accessories for MaxQ™ High Performance Orbital Shakers.