Chemical Identifiers
CAS135459-87-9
IUPAC Namestrontium(2+) bis(2-{5-[bis(carboxymethyl)amino]-2-carboxy-4-cyanothiophen-3-yl}acetate)
Molecular FormulaC24H18N4O16S2Sr
InChI KeyKVJIIORUFXGTFN-UHFFFAOYSA-L
SMILES[Sr++].OC(=O)CN(CC(O)=O)C1=C(C#N)C(CC([O-])=O)=C(S1)C(O)=O.OC(=O)CN(CC(O)=O)C1=C(C#N)C(CC([O-])=O)=C(S1)C(O)=O
View more