Nelfinavir mesylate
Thermo Scientific Chemicals

Nelfinavir mesylate

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4662000101 g
466202500250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466200010
Price (EUR)
1 494,30
キャンペーン価格
1 758,00
Save 263,70 (15%)
Each
-
Add to cart
Quantity:
1 g
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
1 494,30
キャンペーン価格
1 758,00
Save 263,70 (15%)
Each
Add to cart
Chemical Identifiers
CAS159989-65-8
IUPAC Name(3S,4aS,8aS)-N-tert-butyl-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylphenyl)formamido]-4-(phenylsulfanyl)butyl]-decahydroisoquinoline-3-carboxamide; methanesulfonic acid
Molecular FormulaC33H49N3O7S2
InChI KeyNQHXCOAXSHGTIA-SKXNDZRYSA-N
SMILESCS(O)(=O)=O.CC1=C(O)C=CC=C1C(=O)N[C@@H](CSC1=CC=CC=C1)[C@H](O)CN1C[C@H]2CCCC[C@H]2C[C@H]1C(=O)NC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
Specific optical rotation-101° to -115° (20°C, 589 nm) (1% in Methanol) (on dry substance)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0463364Certificate of AnalysisJun 13, 2024466202500, 466200010
A0420115Certificate of AnalysisJan 05, 2023466202500, 466200010
2 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection