Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MRATPLAAP AGSLSRKKRL ELDDNLDTER PVQKRARSGP QPRLPPCLLP LSPPTAPDRA TAVATASRLG PYVLLEPEEG GRAYRALHCP TGTEYTCKVY PVQEALAVLE PYARLPPHKH VARPTEVLAG TQLLYAFFTR THGDMHSLVR SRHRIPEPEA AVLFRQMATA LAHCHQHGLV LRDLKLCRFV FADRERKKLV LENLEDSCVL TGPDDSLWDK HACPAYVGPE ILSSRASYSG KAADVWSLGV ALFTMLAGHY PFQDSEPVLL FGKIRRGAYA LPAGLSAPAR CLVRCLLRRE PAERLTATGI LLHPWLRQDP MPLAPTRSHL WEAAQVVPDG LGLDEAREEE GDREVVLYG (1-358 aa encoded by BC027484 )
A novel inhibitor of cell division, Tribbles, blocks mitosis at a critical point in Drosophila morphogenesis. Recently, mammalian homologues of Tribbles (Trbs) have been identified that control mitogen-activated protein kinase (MAPK) activity. TRIB3 also known as TRB-3, SINK, and SKIP3 is a member of Trbs family. Overexpression of this protein inhibits NF-kB-dependent transcription induced by tumor necrosis factor (TNF) stimulation or its downstream signaling proteins but does not inhibit NF-kB translocation to the nucleus and binding to DNA. TRIB3/SKIP3 is also overexpressed in multiple human tumors and is regulated by hypoxia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DJ1103G7.3; induced in fatty liver dystrophy 2; kinase; Neuronal cell death Inducible Putative Kinase; Neuronal cell death-inducible putative kinase; p65-interacting inhibitor of NF-kappa-B; p65-interacting inhibitor of NF-kappaB; SINK; TRB-3; Tribbles 3; Tribbles homolog 3
Gene Aliases: C20orf97; Ifld2; NIPK; SINK; SKIP3; TRB-3; TRB3; TRIB3
UniProt ID: (Human) Q96RU7, (Mouse) Q8K4K2, (Rat) Q9WTQ6
Entrez Gene ID: (Human) 57761, (Mouse) 228775, (Rat) 246273
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support