Thermo Scientific Chemicals

Desoxyanisoin, 98%, Thermo Scientific Chemicals

Catalog number: A12816.22
100 g, Each
Thermo Scientific Chemicals

Desoxyanisoin, 98%, Thermo Scientific Chemicals

Catalog number: A12816.22
100 g, Each
Quantity
Catalog number: A12816.22
also known as A12816-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
120-44-5
IUPAC Name
1,2-bis(4-methoxyphenyl)ethan-1-one
Molecular Formula
C16H16O3
InChI Key
SICBLYCPRWNHHP-UHFFFAOYSA-N
SMILES
COC1=CC=C(CC(=O)C2=CC=C(OC)C=C2)C=C1
Form
Powder
Assay (GC)
≥97.5%
Appearance (Color)
White to pale cream or pale pink
Melting Point (clear melt)
107.5-113.5?C

Description

Deoxyanisoin react to produce a-bromo-4,4'-dimethoxy-deoxybenzoin, and this reaction could happen in the reagent of CCl4 and bromine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxyanisoin react to produce a-bromo-4,4′-dimethoxy-deoxybenzoin, and this reaction could happen in the reagent of CCl4 and bromine.

Solubility
Sparingly soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text