6-Methylheptane-2,4-dione, is used as a multipurpose intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceutical industry
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Methylheptane-2,4-dione, is used as a multipurpose intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceutical industry
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from Strong oxidizing agents.
RUO – Research Use Only
General References:
- BK Kaymakçıoğlu.; S Rollas. Synthesis, characterization and evaluation of antituberculosis activity of some hydrazones. Il Farmaco. 2002, 57,(7), 595-599.
- P Weyerstahl.; U Splittgerber. Constituents of the leaf essential oil of Hypericum perforatum L. from India. Flavour and fragrance. 1995, 10,(6), 365-370.
- Chelating ligand.