Thermo Scientific Chemicals

1,4-Diiodobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A15335.22
100 g, Each
Thermo Scientific Chemicals

1,4-Diiodobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A15335.22
100 g, Each
Quantity
Catalog number: A15335.22
also known as A15335-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
624-38-4
IUPAC Name
1,4-diiodobenzene
Molecular Formula
C6H4I2
InChI Key
LFMWZTSOMGDDJU-UHFFFAOYSA-N
SMILES
IC1=CC=C(I)C=C1
Appearance (Color)
White or pale cream to cream or pale brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
127-135°C
Form
Powder or crystalline powder
Identification (FTIR)
Conforms

Description

1,4-Diiodobenzene is used in Suzuki reaction. It is used as a precursor in the preparation of martinellic acid and 1,4-bis(p-R-phenylethynyl)benzenes. Further, it is employed in the preparation of 1,4-diiodo-2,5-didodecylbenzene , which is a starting reagent for the synthesis of oligo(1,4-phenylene ethynylene). It is also used in human and animal nutrition products including antiseptics and disinfectants. In addition to this, it used in polarizing films for liquid crystal display chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diiodobenzene is used in Suzuki reaction. It is used as a precursor in the preparation of martinellic acid and 1,4-bis(p-R-phenylethynyl)benzenes. Further, it is employed in the preparation of 1,4-diiodo-2,5-didodecylbenzene , which is a starting reagent for the synthesis of oligo(1,4-phenylene ethynylene). It is also used in human and animal nutrition products including antiseptics and disinfectants. In addition to this, it used in polarizing films for liquid crystal display chemicals.

Solubility
Soluble in ether and ethanol. Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing