Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: EASTVVLLNS TDSSPPTNNF TDIEAALKAQ LDSADIPKAR RKRYISQNDM IAILDYHNQV RGKVFPPAAN MEYMVWDENL AKSAEAWAAT CIWDHGPSYL
This gene encodes a trypsin inhibitor. The protein shares similarity to insect venom allergens, mammalian testis-specific proteins and plant pathogenesis-related proteins. It is frequently expressed in human neuroblastoma and glioblastoma cell lines, and thus may play a role in the central nervous system. [provided by RefSeq].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 25 kDa trypsin inhibitor; CRISP-8; Cysteine-rich secretory protein 8; DKFZp686F0366; p25TI; Peptidase inhibitor 15; PI-15; protease inhibitor 15; SugarCrisp
Gene Aliases: CRISP8; P24TI; P25TI; PI15
UniProt ID: (Human) O43692
Entrez Gene ID: (Human) 51050
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support