Glycine dimethylamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycine dimethylamide, 97%, Thermo Scientific Chemicals
 
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H50293.03
also known as H50293-03
Price (EUR)/ Each
275,40
Online exclusive
306.00 
Save 30,60 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)/ Each
275,40
Online exclusive
306.00 
Save 30,60 (10%)
Add to cart
Glycine dimethylamide, 97%, Thermo Scientific Chemicals
Catalog numberH50293.03
Price (EUR)/ Each
275,40
Online exclusive
306.00 
Save 30,60 (10%)
-
Add to cart
Chemical Identifiers
CAS1857-19-8
IUPAC Name(dimethylcarbamoyl)methanaminium
Molecular FormulaC4H11N2O
InChI KeyKNVRBEGQERGQRP-UHFFFAOYSA-O
SMILESCN(C)C(=O)C[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4740-1.4790 @ 20?C
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Glycine dimethylamide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine dimethylamide is used as pharmaceutical intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents and acids.
RUO – Research Use Only

General References:

  1. D.G.H.Ballard; C.H.Bamford. 65. Stereochemical aspects of the reactions between α-N-carboxy-amino-acid anhydrides and primary and secondary bases. J. Chem. Soc. 1958, 355-360
  2. John D.Glass; I.L.Schwartz; Roderich Walter. Enzymatic inactivation of peptide hormones possessing a C-terminal amide group. Current Issue. 1969, 63, (4),1426-1430