N-Boc-thiourea, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Boc-thiourea, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63599.03
also known as H63599-03
Price (EUR)/ Each
83,04
Online exclusive
93.30 
Save 10,26 (11%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)/ Each
83,04
Online exclusive
93.30 
Save 10,26 (11%)
Add to cart
N-Boc-thiourea, 97%, Thermo Scientific Chemicals
Catalog numberH63599.03
Price (EUR)/ Each
83,04
Online exclusive
93.30 
Save 10,26 (11%)
-
Add to cart
Chemical Identifiers
CAS268551-65-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
N-Boc-thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Silvio Cunha; Maısa B Costa; Hamilton B Napolitano; Carlito Lariucci; Ivo Vencato. Study of N-benzoyl-activation in the HgCl2-promoted guanylation reaction of thioureas. Synthesis and structural analysis of N-benzoyl-guanidines. Tetrahedron. 2001, 57(9), 1671-1675.
  2. Kazuo Nagasawa; Angelina Georgieva; Hiroyuki Koshino; Tadashi Nakata; Tetsuya Kita; and Yuichi Hashimoto. Total Synthesis of Crambescidin 359. Org. Lett. 2002, 4(2), 177-180.