Thermo Scientific Chemicals

N-Fmoc-N-methyl-D-leucine, 97%, Thermo Scientific Chemicals

Catalog number: H63895.06
5 g, Each
Thermo Scientific Chemicals

N-Fmoc-N-methyl-D-leucine, 97%, Thermo Scientific Chemicals

Catalog number: H63895.06
5 g, Each
Quantity
Catalog number: H63895.06
also known as H63895-06
Price (EUR)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103478-63-3
IUPAC Name
(2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-4-methylpentanoic acid
Molecular Formula
C22H25NO4
InChI Key
BUJQSIPFDWLNDC-GNLPSFAGNA-N
SMILES
CC(C)C[C@@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Melting Point
110-120?C
Optical Rotation
+19? to +25? (c=1 in DMF)
Assay (HPLC)
≥96.0%

Description

Tripeptides were prepared by means of coupling of N-Fmoc-N-methyl-D-leucine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tripeptides were prepared by means of coupling of N-Fmoc-N-methyl-D-leucine.

Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text