Thermo Scientific Chemicals

DL-2-Aminoadipic acid, 98%, Thermo Scientific Chemicals

Catalog number: L12717.03
1 g, Each
Thermo Scientific Chemicals

DL-2-Aminoadipic acid, 98%, Thermo Scientific Chemicals

Catalog number: L12717.03
1 g, Each
Quantity
Catalog number: L12717.03
also known as L12717-03
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
542-32-5
Appearance (Color)
White to cream to yellow
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥94.0% to ≤106.0%
Identification (FTIR)
Conforms

Description

DL-2-Aminoadipic acid is a labeled amino acid which is used in chemical research and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-2-Aminoadipic acid is a labeled amino acid which is used in chemical research and as pharmaceutical intermediate.

Solubility
Soluble in water (2.2 mg/ml at 20°C), 1 M HCl (50 mg/ml), ethanol (slightly), ether (slightly), and 80% formic acid (25 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Keep in a dry place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text