Mycoplasma / Ureaplasma Agar
Mycoplasma / Ureaplasma Agar
Thermo Scientific™

Mycoplasma / Ureaplasma Agar

Isolate, differentiate and enumerate Mycoplasma and Ureaplasma spp. with a nutritious media that includes antibiotics which inhibit most interferring bacteria and yeasts.
Have Questions?
Catalog NumberDescriptiondimFormatQuantity
PO5081AMycoplasma / Ureaplasma Agar10/Pk.
Catalog number PO5081A
Price (EUR)
-
Request A Quote
Description:
Mycoplasma / Ureaplasma Agar
Quantity:
10/Pk.
Ask our AI about this Product

Detect, isolate, differentiate and enumerate Mycoplasma and Ureaplasma species mainly from urogenital specimens. Find clarity with an antibiotic mixture in the medium that inhibits most gram-negative and gram-positive bacteria, as well as yeasts, which might be present in patient specimens. On Thermo Scientific™ Mycoplasma / Ureaplasma Agar, the colorless colonies of Mycoplasma hominis form the typical fried egg appearance (growth density-dependent). Colonies of Ureaplasma urealyticum are dark-brown and grow in typical sea urchin morphology.

Mycoplasma and Ureaplasma species are parasites on the surface of human and animal epithelium cells. Because some metabolic pathways are missing they are completely dependent on their hosts which deliver them the essential growth factors. So besides the rich peptone base, the medium includes the necessary nutrients (Vitox, cystein, yeast extract, urea and horse serum) which are in vivo offered by the host. U. urealyticum metabolizes urea which results in a pH shift to alkaline conditions. Therefore, manganese sulphate is oxidised into manganese oxide which is incorporated by U. urealyticum giving the dark brown color to the colonies. The pH shift leads to the color shift of the pH indicator phenol red. As a consequence the medium around the Ureaplasma colonies turns from yellow into red.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
ColorTraffic yellow, transparent
DescriptionMycoplasma / Ureaplasma Agar
Fill Weight13.5g ± 5%
Format90mm Monoplate
Packaging TypeFlow Wrapped - Single Layer
Quantity10/Pk.
Shelf Life6 Weeks
Manufacturing OriginGermany
Product TypeAgar
Unit SizeEach
Contents & Storage
6°C to 12°C

Intended use statement 

A medium which supports the growth of Mycoplasma and Ureaplasma species from genital and respiratory samples.

Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
6283827Certificate of AnalysisJun 24, 2025PO5081A
6270857Certificate of AnalysisJun 14, 2025PO5081A
6260954Certificate of AnalysisJun 14, 2025PO5081A
6258125Certificate of AnalysisJun 14, 2025PO5081A
6268645Certificate of AnalysisJun 14, 2025PO5081A
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection