Thermo Scientific Chemicals

Chlorure de cobalt(II) hexahydraté, Puratronic™, 99,998 % (base métallique), Thermo Scientific Chemicals

Référence: 010692.18
50 g, Each
Thermo Scientific Chemicals

Chlorure de cobalt(II) hexahydraté, Puratronic™, 99,998 % (base métallique), Thermo Scientific Chemicals

Référence: 010692.18
50 g, Each
Quantité
Informational:Informatif:Il est possible que nous ayons besoin d’informations supplémentaires concernant ce produit chimique pour nos rapports de conformité réglementaire et chimique. Nous vous contacterons pour obtenir ces informations une fois votre commande passée, si nécessaire.
Référence: 010692.18
également connu sous le numéro 010692-18
Prix (EUR)
Quantité
-
Have Questions?

Identifiants chimiques

CAS
7791-13-1
IUPAC Name
λ²-cobalt(2+) hexahydrate dichloride
Molecular Formula
Cl2CoH12O6
InChI Key
GFHNAMRJFCEERV-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Total Metal Impurities
0.002% max.

Description

Cobalt(II) chloride hexahydrate has been used to study apoptotic effects in HepG2 cells. Also reported as a stabilizer for reconstitution of the radiopharmaceutical technetium-99m exametazime. It has been used in studies to examine heat shock protein expression of inflamed skin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text

    Sécurité et manipulation


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Respiratory Sensitization
    Category 1
    Skin Sensitization
    Category 1
    Germ Cell Mutagenicity
    Category 2
    Carcinogenicity
    Category 1B
    Reproductive Toxicity
    Category 1B
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 + H332 - Harmful if swallowed or if inhaled

    H317 - May cause an allergic skin reaction

    H334 - May cause allergy or asthma symptoms or breathing difficulties if inhaled

    H341 - Suspected of causing genetic defects

    H350i - May cause cancer by inhalation

    H360F - May damage fertility

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P333 + P313 - If skin irritation or rash occurs: Get medical advice/attention

    Additional EU labelling

    Restricted to professional users