D-Sorbitol, 98, %
Thermo Scientific Chemicals

D-Sorbitol, 98, %

Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
500 g
2 kg
Référence 036404.36
également connu sous le numéro 036404-36
Prix (EUR)/ Each
33,48
Offre exceptionnelle en ligne
37.20 
Économisez 3,72 (10%)
-
Ajouter au panier
Quantité:
500 g
Grand volume ou format personnalisé
Prix (EUR)/ Each
33,48
Offre exceptionnelle en ligne
37.20 
Économisez 3,72 (10%)
Ajouter au panier
D-Sorbitol, 98, %
Référence036404.36
Prix (EUR)/ Each
33,48
Offre exceptionnelle en ligne
37.20 
Économisez 3,72 (10%)
-
Ajouter au panier
Identifiants chimiques
CAS50-70-4
IUPAC Namehexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H14O6
InChI KeyFBPFZTCFMRRESA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(O)CO
Voir plus
Appearance (Color)White
FormPowder or crystalline powder
Assay (Silylated GC)≥97.5%
Residual water≤1.5%
Residue on ignition≤0.1%
D-sorbitol is added to food products as sweetener. It is also used in industrial and medical applications. Sorbitol often is applied in modern cosmetics as a humectant and thickener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only